Learn More
Aviva Systems Biology Corp ACTR1B Antibody - middle region (ARP52098_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52098_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACTR1B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ACTR1B. Alternate names: ARP1B, CTRN2, PC3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-63044