Learn More
Aviva Systems Biology Corp ACTR2 Antibody - N-terminal region (ARP46022_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46022_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ACTR2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ACTR2. Alternate names: ARP2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59181