Learn More
Aviva Systems Biology Corp ACTR3B Antibody - N-terminal region (ARP57420_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57420_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACTR3B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ACTR3B. Alternate names: ARP11, ARP3BETA, DKFZp686O24114ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54048