Learn More
Creative Biomart Inactivated SARS-CoV-2 Vaccine Antigen (Chinese Academy of Medical Sciences)
Catalog No. 502084841
Encompass_Preferred
The SARS-CoV-2 Inactivated Vaccine is a purified vaccine that contains multiple antigens from the virus and is inactivated in Vero Cells1. This vaccine specifically was developed in collaboration with the Chinese Academy of Medical Sciences, West China Second University Hospital, and Yunan Center for Disease Control and Prevention. As of June, 2020, safety and Immunogenicity trials are currently underway for this vaccine (NCT04470609 & NCT04412538).
Catalog No. 50-208-4841 Supplier Creative Biomart Supplier No. THP0328
Add to Cart
Add to Cart
The SARS-CoV-2 Inactivated Vaccine is a purified vaccine that contains multiple antigens from the virus and is inactivated in Vero Cells1. This vaccine specifically was developed in collaboration with the Chinese Academy of Medical Sciences, West China Second University Hospital, and Yunan Center for Disease Control and Prevention. As of June, 2020, safety and Immunogenicity trials are currently underway for this vaccine (NCT04470609 & NCT04412538).