Learn More
Aviva Systems Biology Corp ACTRT1 Antibody - middle region (ARP52580_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52580_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACTRT1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ACTRT1. Alternate names: AIP1, ARIP1, ARP-T1, ARPT1, HSD27, KIAA0705, MGC26590ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60777