Learn More
Aviva Systems Biology Corp ACTRT2 Antibody - C-terminal region (ARP52502_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52502_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACTRT2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ACTRT2. Alternate names: ARPM2, ARPT2, Arp-T2, FLJ25424, HARPM2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-60755