Learn More
Grainger Actuated Ball Valve 1/2 In 316 SS
Catalog No. 502748902
Encompass_Preferred
The FisherbrandTM 502748902 is a polypropylene microcentrifuge tube rack that holds 96 tubes in an 8x12 array. It is made of autoclavable polypropylene and has alphanumeric grid references for easy tube identification. The rack measures 127x86x45 mm (5x3.4x1.8 in.) and features a detachable lid with condensation rings to prevent sample cross-contamination. This microcentrifuge tube rack provides an organized storage solution for 1.5/2.0mL tubes and is suitable for centrifugation and freezing samples. The sturdy one-piece construction allows for long-term use.
Catalog No. 50-274-8902 Supplier Grainger Supplier No. 14N250
The FisherbrandTM 502748902 is a polypropylene microcentrifuge tube rack that holds 96 tubes in an 8x12 array. It is made of autoclavable polypropylene and has alphanumeric grid references for easy tube identification. The rack measures 127x86x45 mm (5x3.4x1.8 in.) and features a detachable lid with condensation rings to prevent sample cross-contamination. This microcentrifuge tube rack provides an organized storage solution for 1.5/2.0mL tubes and is suitable for centrifugation and freezing samples. The sturdy one-piece construction allows for long-term use.