Learn More
Grainger Actuated Ball Valve 1 In 316 SS
Catalog No. 502752427
Encompass_Preferred
The Pneumatically Actuated Two-Way Ball Valve by ASCO is a 1/4" NPT brass body valve with a stainless steel ball and stem. It has a maximum pressure rating of 150 psi and temperature rating of -20 to 180°F. This normally closed valve shifts positions when pressure is applied to the air pilot. It can be used for on/off control in air, water, oil and inert gas applications. The valve shifts positions in less than 50 milliseconds.
Catalog No. 50-275-2427 Supplier Grainger Supplier No. 14N252
The Pneumatically Actuated Two-Way Ball Valve by ASCO is a 1/4" NPT brass body valve with a stainless steel ball and stem. It has a maximum pressure rating of 150 psi and temperature rating of -20 to 180°F. This normally closed valve shifts positions when pressure is applied to the air pilot. It can be used for on/off control in air, water, oil and inert gas applications. The valve shifts positions in less than 50 milliseconds.