Learn More
Grainger Actuated Ball Valve 2-1/2 In 316 SS
Catalog No. 502751342
Encompass_Preferred
The Grainger website offers a variety of ball valves for plumbing applications. These full port ball valves have brass or stainless steel bodies with chrome plated brass balls and PTFE seats. They feature blowout-proof stems, adjustable packing glands, and lever or T-handle operators. The valves are available in 1/4" to 2" sizes with NPT threaded or solder end connections. Maximum pressure ratings range from 400 to 600 psi at temperatures up to 250°F. Grainger ball valves provide reliable on/off control of water, oil, natural gas, steam, air, and inert gases in residential, commercial, and industrial settings.
Catalog No. 50-275-1342 Supplier Grainger Supplier No. 14N255
The Grainger website offers a variety of ball valves for plumbing applications. These full port ball valves have brass or stainless steel bodies with chrome plated brass balls and PTFE seats. They feature blowout-proof stems, adjustable packing glands, and lever or T-handle operators. The valves are available in 1/4" to 2" sizes with NPT threaded or solder end connections. Maximum pressure ratings range from 400 to 600 psi at temperatures up to 250°F. Grainger ball valves provide reliable on/off control of water, oil, natural gas, steam, air, and inert gases in residential, commercial, and industrial settings.