Learn More
Grainger Actuated Ball Valve 2 In 316 SS
Catalog No. 502748914
Encompass_Preferred
The Pneumatically Actuated Two-Way Ball Valve by ASCO is a 1/4 inch NPT brass body valve with a stainless steel ball and stem. It has a maximum pressure rating of 150 psi and temperature rating of -20 to 180 degrees Fahrenheit. This normally closed valve shifts positions when pressure is applied to the air pilot section. It can be used for on/off and diverting applications with air, water, oil and inert gas. The valve shifts positions in less than 50 milliseconds.
Catalog No. 50-274-8914 Supplier Grainger Supplier No. 14N254
The Pneumatically Actuated Two-Way Ball Valve by ASCO is a 1/4 inch NPT brass body valve with a stainless steel ball and stem. It has a maximum pressure rating of 150 psi and temperature rating of -20 to 180 degrees Fahrenheit. This normally closed valve shifts positions when pressure is applied to the air pilot section. It can be used for on/off and diverting applications with air, water, oil and inert gas. The valve shifts positions in less than 50 milliseconds.