Learn More
Grainger Actuated Ball Valve 3 In 316 SS
Catalog No. 502752502
Encompass_Preferred
The electrically actuated ball valves on Grainger's website are designed for on/off control of fluids in industrial applications. They feature a ball and socket design that rotates 90 degrees to stop or start flow. The valves are available in bronze or stainless steel bodies in sizes from 1/4" to 2" NPT. They can be actuated with 24VAC/DC or 120VAC solenoid valves and have a Cv flow coefficient range of 4.5 to 32.0. The valves have a pressure rating up to 600 psi and temperature range of -4 to 257°F. They are manufactured by companies like ASCO and NIBCO.
Catalog No. 50-275-2502 Supplier Grainger Supplier No. 14N256
The electrically actuated ball valves on Grainger's website are designed for on/off control of fluids in industrial applications. They feature a ball and socket design that rotates 90 degrees to stop or start flow. The valves are available in bronze or stainless steel bodies in sizes from 1/4" to 2" NPT. They can be actuated with 24VAC/DC or 120VAC solenoid valves and have a Cv flow coefficient range of 4.5 to 32.0. The valves have a pressure rating up to 600 psi and temperature range of -4 to 257°F. They are manufactured by companies like ASCO and NIBCO.