Learn More
Electron Microscopy Sciences AFM Active Air Isolation Platform Self Leveling, 60 x 120 cm, White Epoxy Surface
SDP
Catalog No. 5034439
Encompass_Preferred
These High Performance anti-vibration platforms for microscopes are self-leveling and ideal for use when loads on the platform are likely to change. They can support up to 160kg and are available in a range of standard sizes from 60x60cm up to 150x90cm. Their low 6cm working height and low weight makes them ideal for use on desks and benches.
Size: 60 x 120 cm
Surface: White Epoxy
: Active Air Self Leveling
Catalog No. 50-344-39 Supplier Electron Microscopy Sciences Supplier No. 6260
These High Performance anti-vibration platforms for microscopes are self-leveling and ideal for use when loads on the platform are likely to change. They can support up to 160kg and are available in a range of standard sizes from 60x60cm up to 150x90cm. Their low 6cm working height and low weight makes them ideal for use on desks and benches.
Size: 60 x 120 cm
Surface: White Epoxy
: Active Air Self Leveling