Learn More
Abcam Recombinant human c-Myc protein (Active), 50UG
Supplier:  Abcam AB16990150UG
Encompass_Preferred
Recombinant human c-Myc protein (Active) is a Human Full Length protein, in the 1 to 454 aa range, expressed in Escherichia coli, with >93% purity, 5 EU/µg endotoxin level and suitable for western blot, SDS-PAGE, Functional studies and mass spectrometry. The predicted molecular weight of ab169901 protein is 53 kDa. - Save time and ensure accurate results - use our recombinant c-Myc protein as a control - Optimal protein bioactivity, stability and reproducibility - Cited in over 10 publications
Protein/ Peptide/ Enzyme Synonyms: BHLHE39, MYC, Myc proto-oncogene protein, Class E basic helix-loop-helix protein 39, Proto-oncogene c-Myc, Transcription factor p64, bHLHe39
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0695582