Learn More
OriGene ACTV RECPTR TYPE IIA (ACVR2A)
SDP
Supplier:  OriGene RC210227
Encompass_Preferred
Activin Receptor Type IIA (ACVR2A) (NM_001616) Human Tagged ORF Clone Expression Plasmid 10 ug | Product Components: The ORF clone is ion-exchange column purified and shipped in a 2D barcoded Matrix tube containing 10ug of transfection-ready, dried plasmid DNA (reconstitute with 100 ul of water).
Catalog No. NC2155050