Learn More
OriGene ACTV RECPTR TYPE IIB (ACVR2B)
SDP
Supplier:  OriGene RC212816
Encompass_Preferred
Activin Receptor Type IIB (ACVR2B) (NM_001106) Human Tagged ORF Clone 10 ug | Product Components: The ORF clone is ion-exchange column purified and shipped in a 2D barcoded Matrix tube containing 10ug of transfection-ready, dried plasmid DNA (reconstitute with 100 ul of water).
Catalog No. NC2155047