Learn More
Electron Microscopy Sciences Activated Carbon Filters, Manufacturer: Leica. Model: Coverslipper CV5030/Tissue Processor TP1020 for Organics. Use: 3 months., 1/EA
SDP
Catalog No. 502090714
Encompass
The carbon filter cartridges contain a high grade activated carbon that effectively absorbs most organic and inorganic vapors, including, but not limited to, xylene, formalin, methanol, toluene, and isopropyl alcohol. The filters are an alternate to brand-name original carbon filters and perform the same, if not better, at a fraction of the cost. The sizing and fit is identical to brand name cartridges. Manufacturer: Leica. Model: Coverslipper CV5030/Tissue Processor TP1020 for Organics . Use: 3 months .
Catalog No. 50-209-0714 Supplier Electron Microscopy Sciences Supplier No. 6265405
The carbon filter cartridges contain a high grade activated carbon that effectively absorbs most organic and inorganic vapors, including, but not limited to, xylene, formalin, methanol, toluene, and isopropyl alcohol. The filters are an alternate to brand-name original carbon filters and perform the same, if not better, at a fraction of the cost. The sizing and fit is identical to brand name cartridges. Manufacturer: Leica. Model: Coverslipper CV5030/Tissue Processor TP1020 for Organics . Use: 3 months .