Learn More
Cayman Chemical ACtyl HexapeptID38trIflu 10mg
SDP
Catalog No. 502938338
Encompass_Preferred
A hexapeptide; decreases IL-8 secretion in human keratinocytes when used alone or in combination with acetyl pentapeptide-46
Catalog No. 50-293-8338 Supplier Cayman Chemical Supplier No. 2725810 MG
A hexapeptide; decreases IL-8 secretion in human keratinocytes when used alone or in combination with acetyl pentapeptide-46