Learn More
Cayman Chemical Acetyl Tetrapeptide-3 (acetate), Purity: ≥95%, 100 mg
SDP
Catalog No. 502937415
Encompass_Preferred
A biomimetic peptide; reases protein levels of type III collagen and total laminins in MRC5 human fibroblasts at 1 μM; reverses corticoid-induced decreases in collagen VII expression at the dermal-epidermal junction in human skin explants
Catalog No. 50-293-7415 Supplier Cayman Chemical Supplier No. 27151100 MG
A biomimetic peptide; reases protein levels of type III collagen and total laminins in MRC5 human fibroblasts at 1 μM; reverses corticoid-induced decreases in collagen VII expression at the dermal-epidermal junction in human skin explants