Learn More
Cayman Chemical ACtylCoA Carboxylase 1 Rabb
SDP
Catalog No. 502948377
Encompass_Preferred
A cGAS inhibitor (IC50 0.11 UM) inhibits dsDNA-induced IFN-B1-dependent gene expression in a reporter assay reduces basal Ifnb1 mRNA expression in BMDMs generated from the Trex1-/- mouse model of Aicardi-Goutières syndrome
Catalog No. 50-294-8377 Supplier Cayman Chemical Supplier No. 32192100 UL
A cGAS inhibitor (IC50 0.11 UM) inhibits dsDNA-induced IFN-B1-dependent gene expression in a reporter assay reduces basal Ifnb1 mRNA expression in BMDMs generated from the Trex1-/- mouse model of Aicardi-Goutières syndrome