Learn More
MSC Acu-Rite Digital Readout System, 3 Axes, 7 inch Color Display, Milling and Turning Functions
Catalog No. 5000516004
Encompass
The DRO203 Digital Readout System enhances machine tool profitability by eliminating manual calculations on up to three axes. It significantly improves accuracy and efficiency in manual machine operations. With pre-programmed common cycles and specialized milling/turning functions, it continuously displays true axis location, leading to higher-quality workpieces, less scrap, and tighter tolerances.
  • Eliminates manual calculations on up to three axes
  • Improves machine tool accuracy and efficiency
  • Pre-programmed common cycles with milling and turning functions
  • Continuously displays true location of all axes
  • 7 inch color display with day/night switching and dynamic zoom
  • Absolute and incremental display modes
  • Designed and built in the USA with rugged die-cast construction
  • Operating temperature range of 0-45° C
Catalog No. 50-005-16004 Supplier MSC Supplier No. 79525127
The DRO203 Digital Readout System enhances machine tool profitability by eliminating manual calculations on up to three axes. It significantly improves accuracy and efficiency in manual machine operations. With pre-programmed common cycles and specialized milling/turning functions, it continuously displays true axis location, leading to higher-quality workpieces, less scrap, and tighter tolerances.
  • Eliminates manual calculations on up to three axes
  • Improves machine tool accuracy and efficiency
  • Pre-programmed common cycles with milling and turning functions
  • Continuously displays true location of all axes
  • 7 inch color display with day/night switching and dynamic zoom
  • Absolute and incremental display modes
  • Designed and built in the USA with rugged die-cast construction
  • Operating temperature range of 0-45° C