Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEKISUI Diagnostics™ Acucy™ Reader System

Catalog No. 22321125 Shop All SEKISUI Diagnostics Products
Change view
Click to view available options
Type:
Accuracy Reader
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
22-321-125 Accuracy Reader
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-321-125 Supplier SEKISUI Diagnostics™ Supplier No. 1039
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Acucy™ Reader System eliminates misdiagnoses from misread test results while getting more flexibility in your environment.

Let the Acucy™ Reader be your eyes in the lab. This diagnostic reader uses a reflectance-based measurement method to evaluate the line signal intensities on the test strip placed inside the tray, and a specific algorithm determines the presence or absence of the target analyte. The result from the test is displayed on the LCD screen as positive, negative, or invalid.

Easy-to-Use

  • Scan the 2-D assay-specific barcode on the cassette leveraging the integrated scanner
  • Place the cassette within the sample tray
  • Place the correct number of drops of solution into the well on the test cassette and close drawer - reader will analyze the results at the appropriate time (when using walk away mode)
  • Results are displayed on the touchscreen LCD

More Flexibility for Your Busy Lab

  • Run individual tests, or read tests in batches
  • Walk Away Mode - no need to monitor and keep time. Test cassette is inserted into the reader, the sample is added to the cassette well and the development and timing of the test occur within the reader, with the reader capturing the result when time is up.
  • Read Now Mode - run and batch multiple patient samples simultaneously. Test cassette development time is tracked by the operator outside of the reader, then the cassettes are inserted into the reader.
  • Monitoring Mode - get positive results faster. While in walk away mode, the monitoring mode can be set to read the cassette every minute of the development time. If a positive result is detected, it will display POS (+) and then will continue to run the test until the assay-specific time limit has been reached.

Results Delivered the Way You Need

  • Screen - test result displayed on the LCD
  • Printed - results can be printed on the Acucy™ printer
  • CSV Data File - each result can be transmitted to a comma-separated values (CSV) file allowing for the import of data to a computer or other external device

The Acucy™ System is made up of the Acucy reader, printer, exclusive USB memory drive, password card, and calibration device.

Use only Acucy tests with the Acucy Reader.
Intended for professional and laboratory use only.

TRUSTED_SUSTAINABILITY

Specifications

Type Accuracy Reader
For Use With (Application) Nasal Swab Testing for Influenza A&B
Operating Temperature 15°C to 30°C (59°F to 86°F)
Includes Acucy Reader, Acucy Printer, Acucy Calibration Device, USB Memory Drive, Power Cords, Power Converters, Password Card, System Manual
Dimensions (D x W x H) 9.25 x 4.7 x 5.7 in.
Depth (English) 9.25 in.
Depth (Metric) 235 mm
Height (English) 5.7 in.
Height (Metric) 145 mm
Width (English) 4.7 in.
Width (Metric) 120 mm
Electrical Requirements 100/240 V, 50/60 Hz
Memory Patient Results: 200, External Quality Control Results: 100, Calibration Results: 200
Weight (English) 3.75 lb.
Weight (Metric) 1.7 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.