Learn More
New England Biolabs, Inc. AcuI – 300 units
SDP
Catalog No. 50812394
Encompass
A restriction endonuclease that recognizes the sequence CTGAAGNNNNNNNNNNNNNN_NN^.
Catalog No. 50-812-394 Supplier New England Biolabs, Inc. Supplier No. R0641S
A restriction endonuclease that recognizes the sequence CTGAAGNNNNNNNNNNNNNN_NN^.