Learn More
Medchemexpress LLC HY-16715 5mg Medchemexpress, Acumapimod CAS:836683-15-9 Purity:>98%
SDP
Catalog No. 502081907
Encompass_Preferred
Medchemexpress, HY-16715 5mg Acumapimod CAS:836683-15-9 Acumapimod (BCT197) is an orally active p38 MAP kinase inhibitor, with an IC50 of less than 1 μM for p38α. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1907 Supplier Medchemexpress LLC Supplier No. HY167155MG
Medchemexpress, HY-16715 5mg Acumapimod CAS:836683-15-9 Acumapimod (BCT197) is an orally active p38 MAP kinase inhibitor, with an IC50 of less than 1 μM for p38α. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.