Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK LIFE SCIENCES™ Acura 12 Channel Pipette, Manual
SDP

Catalog No. 02912670 Shop All DWK Life Sciences Products
Change view
Click to view available options
Volume (Metric):
20 to 200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
02-912-670 20 to 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-912-670 Supplier DWK LIFE SCIENCES™ Supplier No. W810609
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Delivers precise multichannel pipetting across 20 to 200 μL, lightweight with a digital display, 360° volumetric module rotation, Justip™ tip ejector, and swift-set user calibration

  • Lightweight design for comfortable hand fitting during extended use
  • Precision digital display visible at all times for accurate volume reading
  • Justip™ adjustable tip ejector compatible with most standard tips
  • Swift-set user calibration system for reliable, reproducible results
  • 360° rotation of volumetric module for flexible pipetting angles
TRUSTED_SUSTAINABILITY

Specifications

Compatible Tips Justip™ tip ejector
Autoclavable Autoclavable
Color White
Disposable Reusable
For Use With Pipette Tips
Grip Type Ergonomic
Material Plastic
Number of Channels 12
Product Line Acura
Programmability No
Sterility Non-sterile
Type Multichannel Pipette
Volume (Metric) 20 to 200 μL
Display Digital
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.