Learn More
DWK Life Sciences Acura™ 865 Self Refilling Pipette - Vial Holder
SDP
Catalog No. 02915028 Shop All DWK Life Sciences Products
Change view
Click to view available options
Vial Holder
Catalog No.
02-915-028 Vial Holder
1 options
Catalog No. 02-915-028 Supplier DWK Life Sciences Supplier No. W835009
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Hold vials as the feeding source for continuous pipetting using the Acura™ manual 865 Self Refilling Microdispensor Pipetter. Includes the Feed Needle and Luer connector.
The Acura™ manual 865 Self Refilling Microdispensor Pipetter is a hand-held, self-refilling microdispenser intended for repeat distribution of microliter volumes. Carefully selected materials ensure durability and resistance to aggressive media. The instrument combines the easy handling of a regular micropipette with the flexibility of a repeater pipette.
TRUSTED_SUSTAINABILITY
Specifications
Type Vial Holder
For Use With (Equipment) Acura 865 Self Refilling Pipette
Includes Feed needle, Luer connector