Learn More
DWK Life Sciences Acura™ Manual 865 Self Refilling Microdispenser Pipette Accessory, Manifold
Shop All DWK Life Sciences Products
Description
- 8-Channel
- 9mm Spacing
- Luer Lock
- Autoclavable
Specifications
Specifications
| Adjustable Tip Spacing | Yes |
| Autoclavable | Autoclavable |
| Material | Stainless Steel |
| Number of Channels | 8 |
| Type | Manifold |
| For Use With (Equipment) | Manual 865 Self Refilling Microdispenser Pipette |