Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Acura™ Model 835 Pipette Macrotips, 5 mL
SDP

Catalog No. 02912683 Shop All DWK Life Sciences Products
Change view
Click to view available options
Format:
Bulk
Rack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
02-912-683 Bulk
02-912-684 Rack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-912-683 Supplier DWK Life Sciences Supplier No. W851362
Add to Cart
Add to Cart

For use with Acura™ Model 835 single channel macropipettes (0.5-5mL). Available in a 500-tip bulk pack or in a 100-tip rack.

  • Made from high grade, virgin, metal-free polypropylene
  • Wide selection, superior quality
  • Precision injection molding care for air-tight fit
  • Lot-by-lot consistency
  • Bulk or rack availability
  • High-density filter prevents aerosol contamination
  • DNase and RNase-free filter tips
  • Pyrogen- free sterile filter tips
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 5 mL
Clean Claims DNase, RNase, and Pyrogen free
Format Bulk
Quantity 500 Tips
For Use With (Equipment) Socorex Acura Manual 835
Product Line Acura
Material Virgin Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.