Learn More
Description
- Made from high grade, virgin, metal-free polypropylene
- Wide selection, superior quality
- Precision injection molding care for air-tight fit
- Lot-by-lot consistency
- Bulk or rack availability
- High-density filter prevents aerosol contamination
- DNase and RNase-free filter tips
- Pyrogen- free sterile filter tips
Specifications
Specifications
| Volume (Metric) | 5 mL |
| Clean Claims | DNase, RNase, and Pyrogen free |
| Format | Rack |
| Quantity | 100 Tips |
| For Use With (Equipment) | Socorex Acura Manual 835 |
| Product Line | Acura |
| Material | Virgin Polypropylene |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
