Learn More
CSAFE LLC CSafe Global 3L Refrigerated Courier Solution for 2°C to 8°C. Maintains 2°C to 8°C for 12 hours.
Catalog No. NC1723669
Encompass
CSafe Courier solutions are designed for physicians, pharmacists, researchers, pharmaceutical representatives, and other healthcare professionals making last-mile delivery of temperature-sensitive life science products. Maintains temperature for 12 hours.Product Storage Temperature: 2°C to 8°C (36°F to 46°F) | Payload Volume: 3.5 L (216in³) | Internal Payload Dimensions mm (inches): 132 x 132 x 203 (5.2 x 5.2 x 8).
Catalog No. NC1723669 Supplier CSAFE LLC Supplier No. PX3LU28
CSafe Courier solutions are designed for physicians, pharmacists, researchers, pharmaceutical representatives, and other healthcare professionals making last-mile delivery of temperature-sensitive life science products. Maintains temperature for 12 hours.Product Storage Temperature: 2°C to 8°C (36°F to 46°F) | Payload Volume: 3.5 L (216in³) | Internal Payload Dimensions mm (inches): 132 x 132 x 203 (5.2 x 5.2 x 8).