Learn More
Bachem ACV Trifluoroacetate
SDP
Supplier:  Bachem 4028307.0025
Encompass_Preferred
ACV Trifluoroacetate, H-Aad(Cys-D-Val-OH)-OH, 32467-88-2, C14H25N3O6S, Mr 363.44, 25mg. d-(L-a-Aminoadipyl)-L-cysteinyl-D-valine. ACV is the key intermediate in the biosynthesis of all penicillin and cephalosporin antibiotics by eukaryotic and prokaryotic microorganisms.
Catalog No. 50-259-824