Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ACV Tripeptide TFA | 477.45 | 25 MG
SDP

Catalog No. 5000398516
Encompass_Preferred

ACV Tripeptide TFA is a tripeptide that serves as the initial dedicated intermediate in the biosynthetic pathway leading to penicillin and cephalosporin classes of β-lactam natural products in bacteria and fungi. It is nonribosomally synthesized by the ACV synthetase (ACVS) enzyme, encoded by the 11 kb *pchAB* gene, and can be used for the biosynthesis of β-lactam antibiotics. This tripeptide contains the unusual amino acid α-aminoadipic acid (α-AAA), along with L-cysteine and D-valine.

  • Tripeptide
  • Initial intermediate in β-lactam biosynthesis
  • Synthesized by ACV synthetase (ACVS) enzyme
  • Contains α-aminoadipic acid (α-AAA), L-cysteine, and D-valine
  • Molecular weight: 477.45
  • Formula: C16H26F3N3O8S
  • Appearance: Solid

Catalog No. 50-003-98516 Supplier Medchemexpress LLC Supplier No. 25MGHYP3519A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACV Tripeptide TFA is a tripeptide that serves as the initial dedicated intermediate in the biosynthetic pathway leading to penicillin and cephalosporin classes of β-lactam natural products in bacteria and fungi. It is nonribosomally synthesized by the ACV synthetase (ACVS) enzyme, encoded by the 11 kb *pchAB* gene, and can be used for the biosynthesis of β-lactam antibiotics. This tripeptide contains the unusual amino acid α-aminoadipic acid (α-AAA), along with L-cysteine and D-valine.

  • Tripeptide
  • Initial intermediate in β-lactam biosynthesis
  • Synthesized by ACV synthetase (ACVS) enzyme
  • Contains α-aminoadipic acid (α-AAA), L-cysteine, and D-valine
  • Molecular weight: 477.45
  • Formula: C16H26F3N3O8S
  • Appearance: Solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.