Learn More
Cayman Chemical AcVDVADAFCtrIfluoroACTAt 10mg
SDP
Catalog No. 502914939
Encompass_Preferred
A fluorogenic substrate for caspase-2; AFC is released upon enzymatic cleavage by caspase-2 and its fluorescence can be used to quantify caspase-2 activity; ex/em = 400/505 nm, respectively
Catalog No. 50-291-4939 Supplier Cayman Chemical Supplier No. 3735110 MG
A fluorogenic substrate for caspase-2; AFC is released upon enzymatic cleavage by caspase-2 and its fluorescence can be used to quantify caspase-2 activity; ex/em = 400/505 nm, respectively