Learn More
Cayman Chemical AcVDVADpNAtrIfluoroACTAt 2.5mg
SDP
Catalog No. 502938538
Encompass_Preferred
A colorimetric caspase-2 substrate; cleavage of the VDVAD peptide sequence releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of caspase-2 activity; can also be cleaved, with lower efficiency, by caspase-3 and -7
Catalog No. 50-293-8538 Supplier Cayman Chemical Supplier No. 271342.5 MG
A colorimetric caspase-2 substrate; cleavage of the VDVAD peptide sequence releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of caspase-2 activity; can also be cleaved, with lower efficiency, by caspase-3 and -7