Learn More
Cayman Chemical AcVEIDAMCamonIum acetAT 10mg
SDP
Catalog No. 502970092
Encompass_Preferred
A fluorogenic substrate that can be cleaved by caspase-6 and related proteases including caspase-8
Catalog No. 50-297-0092 Supplier Cayman Chemical Supplier No. 1498910 MG
A fluorogenic substrate that can be cleaved by caspase-6 and related proteases including caspase-8