Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ACVR1 [R206H], GST Tag Recombinant Protein

Catalog No. PV6232
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV6232 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PV6232 Supplier Invitrogen™ Supplier No. PV6232
Add to Cart
Add to Cart

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Recombinant human ACVR1 (R206H) (147-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. Validated in LanthaScreen® Kinase Binding Assay Protocols available for simple use and optimized performance Multiple pack sizes available for convenient use Related Product:LanthaScreen® Eu Kinase Binding Assay Protein Form: Recombinant Assay Entry: Biochemical binding/kinetic.

ACVR1 (ALK2) belongs to the transforming growth factor-beta (TGF-beta) superfamily of structurally related signaling proteins which encodes activin A type I receptor. ACVR1 signals a particular transcriptional response in concert with activin type II receptors. Mutations in ACVR1 are associated with fibrodysplasia ossificans progressive (FOP) and the mutations are located in or adjacent to the GS regulatory region or active site of the kinase. The mutations in ACVR1 were predicted by protein structure homology modeling to activate the ACVR1 protein and enhance receptor signaling. Dysregulated ACVR1 kinase activity plays a role in the pathogenesis of FOP.
TRUSTED_SUSTAINABILITY

Specifications

Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 25% glycerol, 150mM NaCl, 0.25mM DTT, 0.1mM PMSF, 0.1mM EDTA, 10mM glutathione and no preservative; pH 7.5
Molecular Weight (g/mol) 67.6 kDa
Name Human ACVR1 [R206H], GST Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Common Name ACVR1 (R206H Mutant)
Protein Length 147-end
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 147-end
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Assay Entry Biochemical binding/kinetic
Kinase Group Serine/Threonine Kinase
Target Entry ACVR1 (ALK2) R206H
Expression System Insect cells
Protein Form Recombinant
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.