Learn More
Aviva Systems Biology Corp ACVR1 Antibody - N-terminal region (ARP45040_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45040_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACVR1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ACVR1. Alternate names: ACTRI, ACVRLK2, ALK2, FOP, SKR1, TSRI, ACVR1AApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58958