Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ACVR1B (ALK4), GST Tag Recombinant Protein

Catalog No. PR7950A
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR7950A 100 μg
PV4312 10 μg
PV4329 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PR7950A Supplier Invitrogen™ Supplier No. PR7950A
Add to Cart
Add to Cart

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Catalytic.

ACVR1B is a serine/threonine kinase involved in signal transduction and cellular communication. Several truncated ACVR1B isoforms have been reported to be exclusively expressed in human pituitary tumors. Mutations of ACVR1B have been observed in pancreatic adenocarcinomas. Type I and II receptors form a stable complex after ligand binding, resulting in phosphorylation of type I receptors by type II receptors. ACVR1B encodes activin A type IB receptor, composed of 11 exons. Alternative splicing and alternative polyadenylation result in 3 fully described transcript variants. The mRNA expression of variants 1, 2, and 3 is confirmed, and a potential fourth variant contains an alternative exon 8 and lacks exons 9 through 11, but its mRNA expression has not been confirmed.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P36896
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 0.5mM EDTA, 2mM DTT, 150mM NaCl, 50% glycerol, 0.02% Triton X-100 and no preservative; pH 7.5
Gene ID (Entrez) 91
Molecular Weight (g/mol) 68.2 kDa
Name Human ACVR1B (ALK4), GST Tag
pH Range 7.5
Purification Method Purified
Quantity 100 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 6820432J04; activin A receptor type 1B; activin A receptor type IB; activin A receptor, type 1B; activin A receptor, type IB; activin A receptor, type II-like kinase 4; activin receptor type IB; Activin receptor type-1B; activin receptor-like kinase 4; ACTRI; ActRIB; ActR-IB; ACVR1A; Acvr1b; ACVRLK2; ACVRLK4; ALK2; Alk4; ALK-4; FOP; Serine/threonine-protein kinase receptor R2; SKR1; SKR2; TSRI
Common Name ACVR1B
Gene Symbol ACVR1B
Kinase Family STKR Kinase Family
Protein Length 150-505
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 150-505
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Kinase Group Ser⁄Thr Kinases: TKL Group
Shipping Condition Dry Ice
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Catalytic
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Signal Transduction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.