Learn More
Aviva Systems Biology Corp ACVR1C Antibody - N-terminal region (ARP52836_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52836_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACVR1C. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ACVR1C. Alternate names: ACVRLK7, ALK7ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60846