Learn More
Carnabio Usa Inc ACVR2A/5UG/CYTOPLASMIC
Supplier:  Carnabio Usa Inc 091315
Encompass
ACVR2A, Cytoplasmic domain, Wild type, Amino Acid 162-513, NP_001607.1, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0321