Learn More
Aviva Systems Biology Corp ACVR2B Antibody - middle region (ARP45042_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45042_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACVR2B. It was validated on Western Blot. ACVR2B,activin A receptor, type IIB,ACTRIIB; ActR-IIB; MGC116908; ACVR2B,activin receptor type-2B,activin receptor type-2BThe immunogen is a synthetic peptide directed towards the middle region of human ACVR2B. Alternate names: ACTRIIB, ActR-IIB, MGC116908, HTX4ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-285-178