Learn More
Medchemexpress LLC ACVR2B Human/Cynomo 500ug
SDP
Catalog No. 5000181852
Encompass_Preferred
ACVR2B is a type II member of the TGF- family of receptor Serine/Threonine kinases ACVR2B binds to activins and growth differentiation factors (GDF) which in turn activate type I receptors activating downstream molecule SMAD2/3[1] ACVR2B Protein Human/Cynomolgus (HEK293 hFc) is produced in HEK293 cells with a C-Terminal hFc-tag
Catalog No. 50-001-81852 Supplier Medchemexpress LLC Supplier No. HYP72813500UG
ACVR2B is a type II member of the TGF- family of receptor Serine/Threonine kinases ACVR2B binds to activins and growth differentiation factors (GDF) which in turn activate type I receptors activating downstream molecule SMAD2/3[1] ACVR2B Protein Human/Cynomolgus (HEK293 hFc) is produced in HEK293 cells with a C-Terminal hFc-tag