Learn More
OriGene ACVR2B Human shRNA Plasmid Kit, 4 Unique Constructs, RFP Reporter, 5 µg Each
SDP
Catalog No. NC3654233
Encompass_Preferred
The OriGene Technologies Activin Receptor Type IIB (ACVR2B) Human shRNA Plasmid Kit is designed for effective gene knockdown studies. This kit includes four unique 29mer shRNA constructs, facilitating robust silencing of ACVR2B, an important regulator in biological processes. Each construct contains a retroviral vector with a Red Fluorescent Protein (RFP) for monitoring transfection efficiency, providing researchers with a reliable tool for functional genomic analysis.
Catalog No. NC3654233 Supplier OriGene Supplier No. TL320254V
The OriGene Technologies Activin Receptor Type IIB (ACVR2B) Human shRNA Plasmid Kit is designed for effective gene knockdown studies. This kit includes four unique 29mer shRNA constructs, facilitating robust silencing of ACVR2B, an important regulator in biological processes. Each construct contains a retroviral vector with a Red Fluorescent Protein (RFP) for monitoring transfection efficiency, providing researchers with a reliable tool for functional genomic analysis.