Learn More
Cayman Chemical Ac-WEAD-pNA (trifluoroacetate salt), Purity: ≥95%, 5 mg
SDP
Catalog No. 502941096
Encompass_Preferred
A colorimetric caspase-1 and caspase-4 substrate; cleavage of the WEAD peptide sequence releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of caspase-1 and caspase-4 activity
Catalog No. 50-294-1096 Supplier Cayman Chemical Supplier No. 274245 MG
A colorimetric caspase-1 and caspase-4 substrate; cleavage of the WEAD peptide sequence releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of caspase-1 and caspase-4 activity