Learn More
Cayman Chemical AcWEHDAFCtrIfluoroACTAte 10mg
SDP
Catalog No. 502970488
Encompass_Preferred
A fluorogenic substrate that can be cleaved specifically by group I caspases
Catalog No. 50-297-0488 Supplier Cayman Chemical Supplier No. 1499210 MG
A fluorogenic substrate that can be cleaved specifically by group I caspases