Learn More
Medchemexpress LLC HY-111791 10mg Medchemexpress, ACY-1083 CAS:1708113-43-2 Purity:>98%
SDP
Catalog No. 501968967
Encompass_Preferred
Medchemexpress, HY-111791 10mg ACY-1083 CAS:1708113-43-2 ACY-1083 is a selective and brain-penetrating HDAC6 inhibitor with an IC50 of 3 nM and is 260-fold more selective for HDAC6 than all other classes of HDAC isoforms. ACY-1083 effectively reverses chemotherapy-induced peripheral neuropathy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8967 Supplier Medchemexpress LLC Supplier No. HY11179110MG
Medchemexpress, HY-111791 10mg ACY-1083 CAS:1708113-43-2 ACY-1083 is a selective and brain-penetrating HDAC6 inhibitor with an IC50 of 3 nM and is 260-fold more selective for HDAC6 than all other classes of HDAC isoforms. ACY-1083 effectively reverses chemotherapy-induced peripheral neuropathy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.