Learn More
TARGETMOL CHEMICALS INC ACY-1083 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T102445MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ACY-1083 is a selective and brain-penetrating HDAC6 inhibitor (IC50 3 nM) and effectively reverses chemotherapy-induced peripheral neuropathy. purity: 99%
Catalog No. 50-258-1744