Learn More
Medchemexpress LLC HY-16026 100mg , ACY-1215 Ricolinostat;Rocilinostat CAS:1316214-52-4 Purity:98%
SDP
Catalog No. 501492801
Encompass_Preferred
Medchemexpress, HY-16026 100mg ACY-1215 Ricolinostat;Rocilinostat CAS:1316214-52-4 Formula:C24H27N5O3 IC50: 5 nM (HDAC6), 58 nM (HDAC1), 48 nM (HDAC2), 51 nM (HDAC3) Purity:98% ACY-1215 is a potent and selective HDAC6 inhibitor, with an enzymatic IC 50 value of 5 nM. ACY-1215 also inhibits HDAC1 , HDAC2 , and HDAC3 with IC 50 of 58, 48, and 51 nM, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2801 Supplier Medchemexpress LLC Supplier No. HY16026100MG
Medchemexpress, HY-16026 100mg ACY-1215 Ricolinostat;Rocilinostat CAS:1316214-52-4 Formula:C24H27N5O3 IC50: 5 nM (HDAC6), 58 nM (HDAC1), 48 nM (HDAC2), 51 nM (HDAC3) Purity:98% ACY-1215 is a potent and selective HDAC6 inhibitor, with an enzymatic IC 50 value of 5 nM. ACY-1215 also inhibits HDAC1 , HDAC2 , and HDAC3 with IC 50 of 58, 48, and 51 nM, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.