Learn More
Medchemexpress LLC Acy-957 | 1609389-52-7 | 99.6% | 429.54 g/mol | C24H23N5OS | 50 MG
SDP
Catalog No. 5000244272
Encompass_Preferred
ACY-957 is an orally active, selective small-molecule inhibitor of histone deacetylase isoforms HDAC1 and HDAC2, used for research into epigenetic regulation. It shows potent activity against HDAC1 and HDAC2, reduced activity against HDAC3, and no measurable inhibition of several class II HDACs. The compound is supplied as a high-purity solid and is available in solution formats for assay use.
  • Potent inhibition of HDAC1 and HDAC2.
  • High selectivity over HDAC3 and class II HDACs.
  • Suitable for in vitro and in vivo formulation protocols.
  • High reported purity for reliable experimental results.
  • Provided as a solid and as ready-to-use DMSO solutions.
  • Documented storage and stability conditions for sample handling.
Catalog No. 50-002-44272 Supplier Medchemexpress LLC Supplier No. HY10400850MG
ACY-957 is an orally active, selective small-molecule inhibitor of histone deacetylase isoforms HDAC1 and HDAC2, used for research into epigenetic regulation. It shows potent activity against HDAC1 and HDAC2, reduced activity against HDAC3, and no measurable inhibition of several class II HDACs. The compound is supplied as a high-purity solid and is available in solution formats for assay use.
  • Potent inhibition of HDAC1 and HDAC2.
  • High selectivity over HDAC3 and class II HDACs.
  • Suitable for in vitro and in vivo formulation protocols.
  • High reported purity for reliable experimental results.
  • Provided as a solid and as ready-to-use DMSO solutions.
  • Documented storage and stability conditions for sample handling.