Learn More
TARGETMOL CHEMICALS INC ACY-957 5MG
SDP
Catalog No. 502581743
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ACY-957 (HDAC Inhibitor C001) is an orally active and selective inhibitor of HDAC1 and HDAC2 (IC50s 7 nM 18 nM and 1300 nM against HDAC1/2/3) and shows no inhibition on HDAC4/5/6/7/8/9. purity: 98%
Catalog No. 50-258-1743 Supplier TARGETMOL CHEMICALS INC Supplier No. T102455MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ACY-957 (HDAC Inhibitor C001) is an orally active and selective inhibitor of HDAC1 and HDAC2 (IC50s 7 nM 18 nM and 1300 nM against HDAC1/2/3) and shows no inhibition on HDAC4/5/6/7/8/9. purity: 98%