Learn More
Medchemexpress LLC HY-104008 5mg Medchemexpress, ACY-957 CAS:1609389-52-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1040085MG
Encompass_Preferred
Medchemexpress, HY-104008 5mg ACY-957 CAS:1609389-52-7 ACY-957 is a selective inhibitor of HDAC1 and HDAC2 , with IC 50 s of 7 nM, 18 nM, and 1300 nM against HDAC1/2/3, respectively, and shows no inhibition on HDAC4/5/6/7/8/9. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2252